7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine, 7-Bromo-3,4-dihydro-4-methyl-2H-1,4-benzoxazine

OR23220-1G
APOSOR23220-1GEA 62.5 EUR
APOSOR23220-1G
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine
Formel: C₉H₁₀BrNO
Molecular Weight: 228,1 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681913
CAS-Nummer: 154264-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR