2-(3-Pyridinyl)-4-thiazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Pyridin-3-yl)thiazole-4-carboxylic acid

OR23221-250MG
APOSOR23221-250MGEA 29 EUR
APOSOR23221-250MG
2-(3-Pyridinyl)-4-thiazolcarbonsäure
2-(3-Pyridinyl)-4-thiazolcarbonsäure
Formel: C₉H₆N₂O₂S
Molecular Weight: 206,22 g/mol
Schmelzpunkt: 283…284 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052304
CAS-Nummer: 39067-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR