4-(1,3-Oxazol-5-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-Oxazolyl)benzoic Acid

OR23227-250MG
APOSOR23227-250MGEA 21.9 EUR
APOSOR23227-250MG
4-(1,3-Oxazol-5-yl)benzoic acid
4-(1,3-Oxazol-5-yl)benzoic acid
Formel: C₁₀H₇NO₃
Molecular Weight: 189,17 g/mol
Schmelzpunkt: 300…301 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682033
CAS-Nummer: 250161-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR