4-Oxazol-5-ylbenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR23228-1G
APOSOR23228-1GEA
0
EUR
APOSOR23228-1G
4-Oxazol-5-ylbenzenesulphonyl chloride
4-Oxazol-5-ylbenzenesulphonyl chloride
Formel:
C₉H₆ClNO₃S Molecular Weight: 243,67 g/mol Schmelzpunkt: 91…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681962 CAS-Nummer: 337508-66-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...