5-(4-Bromphenyl)oxazol

Lieferant: Apollo Scientific
Achtung

OR23230-1G
APOSOR23230-1GEA 21.9 EUR
APOSOR23230-1G
5-(4-Bromphenyl)oxazol
5-(4-Bromophenyl)oxazole
Formel: C₉H₆BrNO
Molecular Weight: 224,0540 g/mol
Schmelzpunkt: 78…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682015
CAS-Nummer: 72571-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR