1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR23237-5G
APOSOR23237-5GEA
122
EUR
APOSOR23237-5G
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 97%
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde
Formel:
C₇H₁₀N₂O Molecular Weight: 138,17 g/mol Siedepunkt: 132 °C/12mmHg Schmelzpunkt: 71…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159623 CAS-Nummer: 2644-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...