2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxo-1,9-diazaspiro[5.5]undec-3-ene, N9-BOC protected

OR2324-1G
APOSOR2324-1GEA 1200 EUR
APOSOR2324-1G
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester 95+%
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester
Formel: C₁₄H₂₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09998740
CAS-Nummer: 1031927-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR