2-Methyl-5-phenylfuran-3-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR23241-1G
APOSOR23241-1GEA
0
EUR
APOSOR23241-1G
2-Methyl-5-phenylfuran-3-carbonyl chloride
2-Methyl-5-phenylfuran-3-carbonyl chloride
Formel:
C₁₂H₉ClO₂ Molecular Weight: 220,652 g/mol Siedepunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221071 CAS-Nummer: 175276-57-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...