4-Bromo-5-methyl-1-phenyl-1H-pyrazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR23249-1G
APOSOR23249-1GEA
0
EUR
APOSOR23249-1G
4-Bromo-5-methyl-1-phenyl-1H-pyrazole 97%
4-Bromo-5-methyl-1-phenyl-1H-pyrazole
Formel:
C₁₀H₉BrN₂ Schmelzpunkt: 37…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681923 CAS-Nummer: 50877-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...