1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxo-1,9-diazaspiro[5.5]undec-3-ene hydrochloride

OR2325-1G
APOSOR2325-1GEA 1350 EUR
APOSOR2325-1G
1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride 95+%
1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride
Formel: C₉H₁₄N₂O·ClH
Molecular Weight: 202,681 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09998741
CAS-Nummer: 1172814-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR