2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoesäure

Lieferant: Apollo Scientific
Achtung

OR23252-1G OR23252-250MG
APOSOR23252-1GEA 139 EUR
APOSOR23252-1G APOSOR23252-250MG
2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoesäure
2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid
Formel: C₁₂H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02682016
CAS-Nummer: 321309-43-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR