4-(1,2,3-Thiadiazol-4-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR23259-1G OR23259-250MG
APOSOR23259-1GEA 0 EUR
APOSOR23259-1G APOSOR23259-250MG
4-(1,2,3-Thiadiazol-4-yl)benzoic acid
4-(1,2,3-Thiadiazol-4-yl)benzoic acid
Formel: C₉H₆N₂O₂S
Schmelzpunkt: 230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085167
CAS-Nummer: 187999-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR