tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Oxo-1,9-diazaspiro[5.5]undecane, N9-BOC protected

OR2326-1G
APOSOR2326-1GEA 1350 EUR
APOSOR2326-1G
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate 95+%
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate
Formel: C₁₄H₂₄N₂O₃
Molecular Weight: 268,36 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998742
CAS-Nummer: 1031927-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR