4-Morpholinobenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR23267-250MG
APOSOR23267-250MGEA 21.9 EUR
APOSOR23267-250MG
4-Morpholinobenzaldehyd
4-Morpholinobenzaldehyd
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,2264 g/mol
Schmelzpunkt: 65…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00735826
CAS-Nummer: 1204-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR