(4-Morpholin-4-yl-phenyl)methanol 98%

Lieferant: Apollo Scientific
Achtung

OR23268-5G
APOSOR23268-5GEA 65.4 EUR
APOSOR23268-5G
(4-Morpholin-4-yl-phenyl)methanol 98%
(4-Morpholin-4-yl-phenyl)methanol
Formel: C₁₁H₁₅NO₂
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01057413
CAS-Nummer: 280556-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR