1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxo-1,9-diazaspiro[5.5]undecane hydrochloride

OR2327-100MG
APOSOR2327-100MGEA 795 EUR
APOSOR2327-100MG
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride 95%
1,9-Diazaspiro[5.5]undecan-2-one hydrochloride
Formel: C₉H₁₆N₂O·ClH
Molecular Weight: 204,697 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09998743
CAS-Nummer: 1171417-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR