6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-(Chlorosulphonyl)-2-(morpholin-4-yl)pyridine, 4-[5-(Chlorosulphonyl)pyridin-2-yl]morpholine
OR23272-1G
APOSOR23272-1GEA
282
EUR
APOSOR23272-1G
6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride 97%
6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride
Formel:
C₉H₁₁ClN₂O₃S Schmelzpunkt: 118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681929 CAS-Nummer: 337508-68-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...