6-(Morpholin-4-yl)nicotinic acid hydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(4-Morpholino)nicotinohydrazide

OR23277-1G OR23277-250MG
APOSOR23277-1GEA 696 EUR
APOSOR23277-1G APOSOR23277-250MG
6-(Morpholin-4-yl)nicotinic acid hydrazide
6-(Morpholin-4-yl)nicotinic acid hydrazide
Formel: C₁₀H₁₄N₄O₂
Schmelzpunkt: 196…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407297
CAS-Nummer: 388088-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR