1,9-Diazaspiro[5.5]undec-3-ene, N9-BOC protected 95+%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 1,9-diazaspiro[5.5]undec-3-ene-9-carboxylate

OR2328-1G
APOSOR2328-1GEA 1420 EUR
APOSOR2328-1G
1,9-Diazaspiro[5.5]undec-3-ene, N9-BOC protected 95+%
1,9-Diazaspiro[5.5]undec-3-ene, N9-BOC protected
Formel: C₁₄H₂₄N₂O₂
Molecular Weight: 252,353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09998744
CAS-Nummer: 1031927-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR