(2-Phenyl-1,3-thiazol-4-yl)methanol

Lieferant: Apollo Scientific
Achtung

OR23285-250MG
APOSOR23285-250MGEA 21.9 EUR
APOSOR23285-250MG
(2-Phenyl-1,3-thiazol-4-yl)methanol
(2-Phenyl-1,3-thiazol-4-yl)methanol
Formel: C₁₀H₉NOS
Schmelzpunkt: 67…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02682017
CAS-Nummer: 23780-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR