(2-Phenyl-1,3-thiazol-4-yl)methylamine
Lieferant: Apollo Scientific
|
Gefahr
|
OR23286-1G
APOSOR23286-1GEA
0
EUR
APOSOR23286-1G
(2-Phenyl-1,3-thiazol-4-yl)methylamine
(2-Phenyl-1,3-thiazol-4-yl)methylamine
Formel:
C₁₀H₁₀N₂S Siedepunkt: 118…122 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02682055 CAS-Nummer: 165736-03-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...