4-(Chloromethyl)-2-phenyl-1,3-thiazole

Lieferant: Apollo Scientific
Gefahr

OR23287-1G OR23287-5G OR23287-10G
APOSOR23287-1GEA 105 EUR
APOSOR23287-1G APOSOR23287-5G APOSOR23287-10G
4-(Chloromethyl)-2-phenyl-1,3-thiazole
4-(Chloromethyl)-2-phenyl-1,3-thiazole
Formel: C₁₀H₈ClNS
Molecular Weight: 209.7 g/mol
Schmelzpunkt: 49…50 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00723503
CAS-Nummer: 4771-31-7
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR