(4-Imidazol-1-ylphenyl)methanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1H-Imidazol-1-yl)benzyl alcohol, (4-Imidazol-1-yl-phenyl)methanol

OR23300-1G
APOSOR23300-1GEA 21.9 EUR
APOSOR23300-1G
(4-Imidazol-1-ylphenyl)methanol 95%
(4-Imidazol-1-ylphenyl)methanol
Formel: C₁₀H₁₀N₂O
Molecular Weight: 174.2000 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682059
CAS-Nummer: 86718-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR