1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide hydrate

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(1H-1,2,4-Triazol-1-yl)benzyl bromide hydrobromide hydrate, 1-(Bromomethyl)-4-(1H-1,2,4-triazol-1-yl)benzene hydrobromide hydrate

OR23306-250MG
APOSOR23306-250MGEA 69.6 EUR
APOSOR23306-250MG
1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide hydrate
1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide hydrate
Formel: C₉H₈BrN₃·BrH·H₂O
Molecular Weight: 337,011 g/mol
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17015228
CAS-Nummer: 1138011-23-0
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR