4-(4-Methyl-1-piperazinyl)benzoesäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Carboxyphenyl)-4-methylpiperazine, 4-(4-Methylpiperazinyl)benzoic acid

OR23308-1G OR23308-250MG OR23308-5G
APOSOR23308-1GEA 24.8 EUR
APOSOR23308-1G APOSOR23308-250MG APOSOR23308-5G
4-(4-Methyl-1-piperazinyl)benzoesäure
4-(4-Methylpiperazin-1-yl)benzoic acid
Formel: C₁₂H₁₆N₂O₂
Schmelzpunkt: 270…272 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD02682063
CAS-Nummer: 86620-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR