(4-(4-Methylpiperazin-1-yl)phenyl)methanol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(4-Methylpiperazin-1-yl)benzyl alcohol, 4-(4-Methylpiperazino)benzyl Alcohol

OR23310-1G
APOSOR23310-1GEA 21.9 EUR
APOSOR23310-1G
(4-(4-Methylpiperazin-1-yl)phenyl)methanol
(4-(4-Methylpiperazin-1-yl)phenyl)methanol
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,2800 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682064
CAS-Nummer: 342405-34-9
UN: 2928
ADR: 6.1+8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR