3-(2-Methylthiazol-4-yl)phenylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(2-Methyl-1,3-thiazol-4-yl)aniline

OR23315-250MG
APOSOR23315-250MGEA 43.5 EUR
APOSOR23315-250MG
3-(2-Methylthiazol-4-yl)phenylamine
3-(2-Methylthiazol-4-yl)phenylamine
Formel: C₁₀H₁₀N₂S
Molecular Weight: 190,27 g/mol
Schmelzpunkt: 102…106 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02579873
CAS-Nummer: 89250-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR