6-Phenoxypyridine-3-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR23318-250MG
APOSOR23318-250MGEA
0
EUR
APOSOR23318-250MG
6-Phenoxypyridine-3-sulphonyl chloride
6-Phenoxypyridine-3-sulphonyl chloride
Formel:
C₁₁H₈ClNO₃S Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681949 CAS-Nummer: 368869-91-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...