5-Iodo-2-phenoxypyridine

Lieferant: Apollo Scientific
Achtung

OR23322-1G OR23322-10G
APOSOR23322-1GEA 151 EUR
APOSOR23322-1G APOSOR23322-10G
5-Iodo-2-phenoxypyridine
5-Iodo-2-phenoxypyridine
Formel: C₁₁H₈INO
Molecular Weight: 297.092 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02681952
CAS-Nummer: 352018-92-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR