4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid 97%

Lieferant: Apollo Scientific

OR23325-5G
APOSOR23325-5GEA 0 EUR
APOSOR23325-5G
4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid 97%
4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid
Formel: C₉H₇N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00111662
CAS-Nummer: 216959-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR