5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine

Lieferant: Apollo Scientific
Achtung

OR23333-100MG
APOSOR23333-100MGEA 109 EUR
APOSOR23333-100MG
5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine
5-(Hydroxymethyl)-4-methyl-2-phenylpyrimidine
Formel: C₁₂H₁₂N₂O
Schmelzpunkt: 84,5…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682074
CAS-Nummer: 342405-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR