3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR23335-5G
APOSOR23335-5GEA 0 EUR
APOSOR23335-5G
3-Methyl-5-phenylisoxazole-4-carboxylic acid 97%
3-Methyl-5-phenylisoxazole-4-carboxylic acid
Formel: C11H9NO3
Molecular Weight: 203,1941 g/mol
Schmelzpunkt: 190…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086120
CAS-Nummer: 17153-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR