4-(Brommethyl)-3-methyl-5-phenylisoxazol

Lieferant: Apollo Scientific
Gefahr

OR23339-100MG
APOSOR23339-100MGEA 69.6 EUR
APOSOR23339-100MG
4-(Brommethyl)-3-methyl-5-phenylisoxazol
4-(Bromomethyl)-3-methyl-5-phenylisoxazole
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,1100 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086124
CAS-Nummer: 113841-59-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR