2-Iodbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
OR2334-5G
APOSOR2334-5GEA
21.9
EUR
APOSOR2334-5G
2-Iodbenzylalkohol
2-Iodbenzylalkohol
Formel:
C₇H₇IO Molecular Weight: 234.04 g/mol Siedepunkt: 145 Schmelzpunkt: 89…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004610 CAS-Nummer: 5159-41-1 EINECS: 225-933-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...