[3-(1H-Pyrrol-1-yl)phenyl]methanol

Lieferant: Apollo Scientific

OR23351-1G
APOSOR23351-1GEA 84.1 EUR
APOSOR23351-1G
[3-(1H-Pyrrol-1-yl)phenyl]methanol
[3-(1H-Pyrrol-1-yl)phenyl]methanol
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD03086142
CAS-Nummer: 83140-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR