1-(3-Bromphenyl)pyrrol

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(3-Bromphenyl)pyrrol

OR23352-250MG
APOSOR23352-250MGEA 29 EUR
APOSOR23352-250MG
1-(3-Bromphenyl)pyrrol
1-(3-Bromophenyl)pyrrole
Formel: C₁₀H₈BrN
Molecular Weight: 222,08 g/mol
Schmelzpunkt: 64…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02665243
CAS-Nummer: 107302-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR