3-(1H-Pyrrol-1-yl)benzylamine
Lieferant: Apollo Scientific
|
Gefahr
|
OR23353-1G
APOSOR23353-1GEA
451
EUR
APOSOR23353-1G
3-(1H-Pyrrol-1-yl)benzylamine
3-(1H-Pyrrol-1-yl)benzylamine
Formel:
C₁₁H₁₂N₂ Siedepunkt: 138 °C/1,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086143 CAS-Nummer: 368869-95-8 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...