1,3-Dimethyl-5-(hydroxymethyl)-1H-thieno[2,3-c]pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1,3-Dimethyl-1H-thieno[2,3-c]pyrazol-5-yl)methanol
OR23363-1G
APOSOR23363-1GEA
223
EUR
APOSOR23363-1G
1,3-Dimethyl-5-(hydroxymethyl)-1H-thieno[2,3-c]pyrazole
1,3-Dimethyl-5-(hydroxymethyl)-1H-thieno[2,3-c]pyrazole
Formel:
C₈H₁₀N₂OS Molecular Weight: 182,243 g/mol Schmelzpunkt: 133…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086155 CAS-Nummer: 423769-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...