Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR23367-1G
APOSOR23367-1GEA 119 EUR
APOSOR23367-1G
Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate 97%
Ethyl-4-bromo-3,5-dimethyl-1H-pyrrole-2-carboxylate
Formel: C₉H₁₂BrNO₂
Molecular Weight: 246,11 g/mol
Schmelzpunkt: 142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02647051
CAS-Nummer: 5408-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR