1-Methyl-1H-benzotriazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-benzo[d][1,2,3]triazole-5-carboxylic acid

OR23371-250MG
APOSOR23371-250MGEA 71.2 EUR
APOSOR23371-250MG
1-Methyl-1H-benzotriazole-5-carboxylic acid
1-Methyl-1H-benzotriazole-5-carboxylic acid
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Schmelzpunkt: 250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086163
CAS-Nummer: 305381-67-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR