2H-Chromene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2H-1-Benzopyran-3-carboxylic acid
OR23377-250MG
APOSOR23377-250MGEA
21.9
EUR
APOSOR23377-250MG
2H-Chromene-3-carboxylic acid
2H-Chromene-3-carboxylic acid
Formel:
C₁₀H₈O₃ Molecular Weight: 176,17 g/mol Schmelzpunkt: 187…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202020 CAS-Nummer: 22649-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...