2H-1-Benzopyran-3-thiocarboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2H-Chromene-3-carbothioic acid amide, 2H-Chromene-3-carbothioamide, 3-Carbamothioyl-2H-1-benzopyran

OR23381-1G
APOSOR23381-1GEA 162 EUR
APOSOR23381-1G
2H-1-Benzopyran-3-thiocarboxamide
2H-1-Benzopyran-3-thiocarboxamide
Formel: C₁₀H₉NOS
Molecular Weight: 191,25 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086175
CAS-Nummer: 423768-57-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR