(2-Morpholinophenyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Morpholin-4-yl-phenyl)methanol
OR23384-250MG
APOSOR23384-250MGEA
30.6
EUR
APOSOR23384-250MG
(2-Morpholinophenyl)methanol
(2-Morpholinophenyl)methanol
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,240 g/mol Schmelzpunkt: 54…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086181 CAS-Nummer: 465514-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...