1-Phenyl-1H-pyrazol-5-carbaldehyd

Lieferant: Apollo Scientific
Achtung

OR23388-250MG
APOSOR23388-250MGEA 49.3 EUR
APOSOR23388-250MG
1-Phenyl-1H-pyrazol-5-carbaldehyd
1-Phenyl-1H-pyrazol-5-carbaldehyd
Formel: C₁₀H₈N₂O
Molecular Weight: 172,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00103497
CAS-Nummer: 132274-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR