3,4-Dimethoxyphenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR2339-25G
APOSOR2339-25GEA
159
EUR
APOSOR2339-25G
3,4-Dimethoxyphenol 97%
3,4-Dimethoxyphenol
Formel:
C₈H₁₀O₃ Molecular Weight: 154.17 g/mol Siedepunkt: 167 °C/14mmHg Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008390 CAS-Nummer: 2033-89-8 EINECS: 217-995-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...