2-(Morpholin-4-yl)pyridine-3-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(Chlorosulphonyl)-2-(morpholin-4-yl)pyridine, 4-[3-(Chlorosulphonyl)pyridin-2-yl]morpholine

OR23390-100MG
APOSOR23390-100MGEA 163 EUR
APOSOR23390-100MG
2-(Morpholin-4-yl)pyridine-3-sulphonyl chloride
2-(Morpholin-4-yl)pyridine-3-sulphonyl chloride
Formel: C₉H₁₁ClN₂O₃S
Molecular Weight: 262,713 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03086194
CAS-Nummer: 465514-17-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR