(2-Morpholino-3-pyridinyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[2-(Morpholin-4-yl)pyridin-3-yl]methanol
OR23392-100MG
APOSOR23392-100MGEA
21.9
EUR
APOSOR23392-100MG
(2-Morpholino-3-pyridinyl)methanol
(2-Morpholino-3-pyridinyl)methanol
Formel:
C₁₀H₁₄N₂O₂ Schmelzpunkt: 78…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086196 CAS-Nummer: 423768-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...