3-(Chlorocarbonyl)piperidine, N-CBZ protected 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-[(Benzyloxy)carbonyl]-3-(chloroformyl)piperidine, Benzyl 3-(chlorocarbonyl)piperidine-1-carboxylate
OR23395-1G
APOSOR23395-1GEA
192
EUR
APOSOR23395-1G
3-(Chlorocarbonyl)piperidine, N-CBZ protected 97%
3-(Chlorocarbonyl)piperidine, N-CBZ protected
Formel:
C₁₄H₁₆ClNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407318 CAS-Nummer: 216502-94-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...