3-Bromimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR23399-1G
APOSOR23399-1GEA
36.4
EUR
APOSOR23399-1G
3-Bromimidazo[1,2-a]pyridin
3-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014556 CAS-Nummer: 4926-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...