1-(Benzofuran-3-yl)-2-bromoethan-1-one
Lieferant: Apollo Scientific
|
Gefahr
|
OR23402-1G
OR23402-250MG
APOSOR23402-1GEA
91.5
EUR
APOSOR23402-1G
APOSOR23402-250MG
1-(Benzofuran-3-yl)-2-bromoethan-1-one
1-(Benzofuran-3-yl)-2-bromoethan-1-one
Formel:
C₁₀H₇BrO₂ Schmelzpunkt: 136…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407369 CAS-Nummer: 187657-92-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...